A propos de cet article
Available in Standard TriView, Slim TriView, Glow Standard TriView, and Glow Slim TriView.
- UL Validated for recycled content
- Made in USA
- High quality
Standard 3-Sided TriView Sign communicates critical safety messages with 180-degree view that significantly increases visibility of information
Standard Glow in the Dark 3-Sided TriView communicates critical safety messages with 180-degree view that significantly increases visibility of information. Glow in the dark feature exceeds 10 hours to meet ASTME 2072-00 standards. Sign is made from high quality 70% post-consumer recycled plastic. Sign measures 7.5"H x 20"W, Projects 5 Inches and includes 2 mounting holes and adhesive tabs. Made in the USA.
ZING Slim 3-Sided TriView Sign communicates critical safety messages with 180-degree view that significantly increases visibility of information. Sign is made from high quality 70% post-consumer recycled plastic. Sign measures 12"H x 9"W, Projects 3 Inches and includes 2 mounting holes and adhesive tabs. Made in the USA.
ZING Slim Glow in the Dark 3-Sided TriView Sign communicates critical safety messages with 180-degree view that significantly increases visibility of information. Glow in the dark feature exceeds 10 hours to meet ASTME 2072-00 standards. Sign is made from high quality 70% post-consumer recycled plastic. Sign measures 12"H x 9"W, Projects 3 Inches and includes 2 mounting holes and adhesive tabs. Made in the USA.
Spécifications
- Catalog No:
- 76025-990
- 76026-038
- 76025-982
- 76025-984
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified
- UL Certified
- UL Certified