A propos de cet article
Adhesive labels are available in three sizes, 7" x 5", 10" x 7", and 14" x 10" with 5/Pack. Triview signs are 1/Pack.
- UL Validated for recycled content
- Made in USA
- High quality
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears, and UV exposure
Three-sided Tri-View Sign offers a 180-degree view giving improved visibility. Sign is made from high quality 70% post-consumer recycled plastic. Sign includes pre-drilled holes and self-adhesive pads for easy mounting and measures 7-1/2"H x 20"W and projects 5 inches. Made in the USA.
The signs adhere directly to walls, recycling bins, and waste receptacles. All Zing signs are made in the USA from recycled materials, which supports sustainability initiatives and LEED credits. The signs are sold 5/pack.
Spécifications
- Catalog No:
- 76023-422
- 76023-424
- 76024-420
- 76022-588
- 76022-612
- 76025-834
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified