ZING Green Safety Eco Safety Sign, DANGER Electrical Equipment Authorized Personnel Only
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
- UL Validated for recycled content
- Made in USA
- High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Certifications: OSHA 29 CFR 1910.145
Spécifications
- Catalog No:
- 76024-708
- 76023-624
- 76023-626
- 76024-704
- 76023-628
- 76024-706
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified