Compact Tube Furnaces, Model R, Nabertherm
Fournisseur: NABERTHERM INC.
Certificats
A propos de cet article
These compact tube furnaces with integrated control systems can be used universally for many processes.
- Single-zoned design as standard; Three-zoned design extends the constant temperature accuracy within the heated length
- Dual shell housing made of sheets of textured stainless steel
- Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
- Tmax 1200 °C: Type N thermocouple, Tmax 1300 °C: Type S thermocouple
- Solid state relays provide for low noise operation
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Equipped with a standard working tube of C530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
BodyTextAccessories: Optional accessories include: Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; charge control with temperature measurement in the working tube and in the furnace chamber outside the tube; Three-zoned design (heated length from 500 mm); variety of working tubes and materials; gas supply systems for protective gas or vacuum operation; process control and documentation via VCD software package for monitoring, documentation and control. Contact your Avantor representative for more information.
Ordering information: These tube furnaces can be ordered in a variety of voltages, temperatures, heated lengths, Single or Three-zone control, and diameters depending on the required charge size. Select Single-zoned or Three-zoned models depending on required temperature uniformity across the needed heated length. In general, Three-zoned models extend the accurate constant temperature in the heated length from one-third of the heated length to one-half. Models are rated up to 1200 or 1300 °C. Control options include the B510 controller (5 programs with 4 segments each), C550 controller (10 programs with 20 segments each), or P580 controller (50 programs with 40 segments each).
Customers Who Bought This Also Bought