SP Wilmad-LabGlass Gas Washing Bottles with Fritted Disc and [ST] 45/50 Top Joint, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Generally used to saturate or dry gas streams, these borosilicate glass gas washing bottles have standard taper 45/50 top joint and stopper component containing gas inlet and outlet tubes.
The lower inlet tube has a coarse (C) disc sealed on to provide a stream of fine bubbles into the bottle.
Complete units are supplied with an 14201-324 plastic clamp.
Customers Who Bought This Also Bought