SHOWA 8814 Heat Resistant Glove, Showa
Fournisseur: Showa
Certificats
A propos de cet article
The SHOWA 8814 heat-resistant gloves are designed to withstand temperatures up to 500°F/260°C. Even after multiple washings, the 8814 maintains its heat-protective qualities.
- The textured surface of the neoprene coating provides superior tactile dexterity
- Easy donning and doffing makes the glove easy to remove in an emergency
- Machine washable
- Rough grip, spray coated neoprene
Gloves are used in high heat applications. Use when handling castings, glassware, or bakeware.
Spécifications
- Catalog No:
- 89022-349
- 89022-343
- 89022-345
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Cotton/Polyester
- Cotton/Polyester
- Cotton/Polyester
- Finish:
- Textured surface of neoprene coating
- Textured surface of neoprene coating
- Textured surface of neoprene coating
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet