Ashing Furnaces with Fold Down or Lift Up Door, Models LV and LVT, Nabertherm
Fournisseur: NABERTHERM INC.
Certificats
A propos de cet article
The ashing furnaces LV 3/11 - LVT 15/11 are especially designed for ashing in the laboratory.
- Maximum temperature: 1100 °C, heating from two sides
- Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
- Insulation made of non-classified fiber material, solid state relays provide for low noise operation
- Housing made of sheets of textured stainless steel, dual shell housing for low external temperatures and stability
- Optional fold down door (LV models) which can be used as work platform or lift up door (LVT models) with hot surface facing away from the operator
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
A special air intake and exhaust system allows air exchange of more than 6 times per minute. Incoming air is preheated to ensure a good temperature uniformity.
BodyTextAccessories: Optional accessories include: Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; process control and documentation via VCD software package for monitoring, documentation, and control. Contact your Avantor representative for more information.
Ordering information: These ashing furnaces can be ordered in a variety of sizes and voltage options, with fold down door (L) or lift up door (LT). All models are rated up to 1100 °C maximum temperature. Control options include the B510 controller (5 programs with 4 segments each) or C550 contrller (10 programs with 20 segments each).
Customers Who Bought This Also Bought