Avantor® Prevail C8, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
- Stable from highly organic to highly aqueous
- Specialty phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
- USP L7
Stable C8 phase. Use for highly hydrophobic compounds that retain too strongly on C18.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
BodyTextAccessories: Use guard cartridges with holder (80101/N) and coupler (HI-081).
Customers Who Bought This Also Bought