Avantor® Prevail C18, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
- Stable from highly organic to highly aqueous
- Specialty phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
- USP L1
Flexibility to switch between varied mobile phase conditions to suit a variety of applications. Excellent sensitivity for microbore applications.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
BodyTextAccessories: Use guard cartridges with holder (80101/N) and coupler (HI-081).
Spécifications
- Catalog No:
- 75797-770
- 75798-922
- 75797-768
- 75798-924
- 75797-774
- 75797-772
- 75798-920
- 75798-930
- 75797-776
- 75798-926
- 75797-780
- 75798-928
- 75797-764
- 75797-762
- 75830-682
- 75830-684
- 75797-760
- 75797-758
- 75812-874
- 75812-872
- 75798-914
- 75830-704
- 75798-916
- 75797-766
- 75813-100
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1