Vydac 202TP C18-HL, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
- USP L1
Higher carbon load than 201TP. Developed for the separation of PAHs.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.
Spécifications
- Catalog No:
- 76398-030
- 75798-288
- Carbon load:
- 9%
- 9%
- Endcapped:
- No
- No
- Surface area:
- 60 - 90 m²/g
- 60 - 90 m²/g
- USP code:
- L1
- L1