VWR® Cell Slide
Supplier: VWR International
Certificates
About this item
The handle of the Cell Slide tilts, allowing operators the convience of direct clamping
- Manufactured from unbreakable, high strength PET
- Only the surface of the Cell Slide has been treated to promote cell adherence; other parts of the device are designed to deter cell adherence
- 2 sizes are avialable: 8 mm and 10 mm
- 12, 18, 32, or 45 Cell Slides are available on both the 8 mm and 10 mm models
- High transparency and optimal light transmittance allow observation under a light microscope or fluorescence microscope
- Set structure supports the development of multi-factor, multi-index, and multi-level in vitro study; resulting in accurate and reliable research results
DNase- and RNase-free. Non-pyrogenic.
The handle is also engraved with numbers to facilitate identification. The integrated Cell Slides are fixed in the culture dish, and if further testing is necessary the Cell Slides can be individually removed and placed in a 48-Well plate or similar container.
Certifications: ISO13485:2003 and ISO9001:2008.
Customers Who Bought This Also Bought