Vydac 238TP C18, HPLC Columns
Fournisseur: Avantor
Certificats
A propos de cet article
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
- USP L1
First generation monomeric C18 phase. Recommended for the same applications as 218TP but offers alternative C18 selectivity.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.
Customers Who Bought This Also Bought