Vydac Everest C18, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Everest columns have unique selectivity and sensitivity, which are the result of bonding technology that improves C18 surface coverage and deactivates residual silanols.
- Unique selectivity for hydrophilic and hydrophobic peptides
- High resolution for complex peptide digests
- Excellent sensitivity with little or no TFA in mobile phase
- USP L1
Vydac 238EV (C18) offers maximum surface coverage for the highest resolution of complex samples. It is recommended for the analysis of complex enzymatic digests (>12 proteins).
Previously, the best 300 Å C18 chemistries have had carbon coverage in the 2.8 to 3.6 µmoll/m² range. Everest C18 coverage is in excess of 4 µmol/m² and approximates the theoretical limit based on surface area. The increased shielding of the base silica increases column lifetime and reduces the amount of TFA required to shield the silica.
Vydac is a registered trademark of W.R.Grace and Co.
Spécifications
- Catalog No:
- 75797-968
- 75799-050
- 75797-972
- 75797-970
- 75799-056
- 75798-688
- 75812-966
- 75798-690
- 75812-968
- 75799-052
- 75799-054
- 75831-448
- 75798-698
- 75798-692
- 75799-030
- 75798-694
- 75798-704
- 75798-700
- 75798-702
- 75799-034
- 75799-036
- 75799-046
- 75799-048
- 75799-042
- 75799-044
- 75830-788
- 75830-668
- 75799-032
- Carbon load:
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1