A propos de cet article
FG-7000 Series Data logging, Digital Force Gauge, Loaded with functionality to simplify compression and tension testing needs. Menu programming for easy selection and set-up of the instrument to desired requirements with three unique modes of operation.
- Three mode operation: Track, Peak and Auto-Peak
- 180° Auto-reversible display, tension/compression icons, and dual printed keypad allows usage as a portable gauge or mounted upside down on a test stand
- Programmable High and Low Limits allow for instant visual and audible indication of pass or fail testing
- Multi-language and engineering unit’s selection provides flexible user comfort
- 1000 point data collection capability can be viewed on display, sent to optional printer, or uploaded to software package for further analysis
The FG-7000 Series digital force gauges are loaded with functionality to simplify your compression and tension testing needs
These high-tech instruments can easily data log a reading at the push of a button for simple data acquisition, or be set to continuous data storage. Data can be viewed on the screen, sent to the optional printer, or loaded to be analyzed and graphed on the free software program. The 1,000 point memory with definable groups allows for multiple tests to be recorded and easily separated upon loading.
The FG-7000’s robust housings are designed to fit perfectly in the operator’s hand for portable testing. The large backlit, 180° auto-reversible display, compression/tension icons, combined with the dual labeled key pad allows for instant switching from push to pull testing for portable or test stand applications. Models are available from 0.5 lbs to 220 lb ranges providing a large offering for various applications such as, incoming quality inspection, finished goods testing, R&D or almost any portable or force stand testing requirement.
The multiple-language FG-7000’s provide menu programming for easy selection and set-up of the instrument to your desired requirements. Three modes of operation are selectable: Track mode for live readings, Peak mode for displaying the peak reading that remains until a higher peak is sensed, and Auto Peak mode which is similar to Peak mode except the peak on the display will additionally reset after a programmed time period. Programmable limits provide a quick visual and audible indication if a test passed or failed on the LCD. In addition, the comparator output can be set up for integration of the instrument into your quality system for repetitive testing such as on a production line.
Spécifications
- Catalog No:
- 75854-074
- 75854-070
- 75854-072
- 75854-066
- 75854-068
- 75854-064
- 75854-060
- 75854-062
- Accuracy:
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- ± 0.2% F.S. accuracy
- Data storage:
- 1000 data points (max.)
- 1000 data points (max.)
- 1000 data points (max.)
- 1000 data points (max.)
- 1000 data points (max.)
- 1000 data points (max.)
- 1000 data points (max.)
- 1000 data points (max.)
- Outputs:
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- USB, Serial Port RS-232, High and Low Limit NPN
- Overload capacity:
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity
- 150% of F.S. Overload Capacity