To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Yeast Media Broth
- Cat. no.:CA101320-684
- Volume:1 L
- Supplier No.:T2740
Spécifications
A propos de cet article
Yeast Media Broth.
- 1 Liter
- Sterile
Contains 2.0% Glucose, 1.0% Casamino acids, 1.06% Ammonium sulfate, 0.74% Monosodium Phosphate, 0.54% Disodium phosphate, 0.34% Yeast Nitrogen Base (-AA/AS), pH 5.6. Ambient shipping conditions. Store at room temperature.