ContactUsLinkComponent
Ward's® Tryptic Soy Agar Dehydrated Media, Powdered, IPM Scientific
Ward's® Tryptic Soy Agar Dehydrated Media, Powdered, IPM Scientific
Catalog # 470202-838
CAS Number:  
WARD'S Tryptic Soy Agar 100g
Ward's® Tryptic Soy Agar Dehydrated Media, Powdered, IPM Scientific
Catalog # 470202-838
CAS Number:  
Specifications
  • Pack type:
    Bag
  • Description:
    Tryptic Soy Agar
  • Size:
    100 g
  • Cat. no.:
    470202-838
  • Supplier No.:
    38-1009
Specifications
About this item
Dilute quantities to any volume as needed.
  • Economical alternative
  • Stores much longer than prepared media
  • Prepare only as much as you need at one time
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.
Customers Who Bought This Also Bought