Ward's® Tryptic Soy Agar Dehydrated Media, Powdered, IPM Scientific
# de catalogue 470024-766
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Bottle
- Description:Tryptic Soy Agar
- Size:500 g
- Cat. no.:470024-766
- Supplier No.:95021-622
Spécifications
A propos de cet article
Dilute quantities to any volume as needed.
- Economical alternative
- Stores much longer than prepared media
- Prepare only as much as you need at one time
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.
Customers Who Bought This Also Bought