Mudcracks
# de catalogue 470226-730
Fournisseur: Avantor
Spécifications
- Length:6"
- Width:10"
- Description:Mudcracks in Argillite
- Cat. no.:470226-730
Spécifications
A propos de cet article
Slab from an ancient mudflat showing filled desiccation cracks.
Customers Who Bought This Also Bought