Accessories for Force EXP Guard Column Cartridges, Restek
# de catalogue 10118-320
Fournisseur: Restek
Spécifications
- Description:EXP Direct Connect Holder for EXP Guard Cartridges, Includes Fitting and Ferrules, Max. pressure: 20,000 psi (1,400 bar)
- Cat. no.:10118-320
- Supplier No.:25808