Vydac Denali C18, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Vydac Denali (238DE) is a 120 Å C18 bonded phase with high carbon coverage, suitable for the analysis of both acidic and basic analytes
- High retentivity
- LC-MS of small molecules
- Fully scalable from capillary to process
- USP L1
- Stainless steel column hardware
It has applications for small molecule analyses of interest to pharmaceutical and environmental laboratories.
Vydac is a registered trademark of W.R.Grace and Co.
Customers Who Bought This Also Bought