Vinyl-Coated Lead Rings, Argos Technologies
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Perfect for stabilizing glassware and laboratory containers.
- Chemical-resistant vinyl coating
- Available in O-shaped or C-shaped rings
- Optional holder for benchtop organization
Lead Ring Weights are ideal for stabilizing glassware and laboratory containers during water immersion. The vinyl coating prevents scratching on labware or the benchtop. The “C” shaped rings have some flexibility for a more customized fit.
Customers Who Bought This Also Bought