Luer Lock Valve Adaptors, Restek
# de catalogue 10862-648
Fournisseur: Restek
Spécifications
- Description:Luer Lock Valve Adaptor for Syringes
- Cat. no.:10862-648
- Supplier No.:21283
Spécifications
A propos de cet article
Use a luer lock valve adaptor to connect a luer lock fitting to a Rheodyne or Valco valve.
Customers Who Bought This Also Bought