SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc and Vacuum Adapter, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
These Buchner filter funnels with vacuum adapter feature a full length Standard Taper inner joint sealed to the stem with side hose connection just above the grinding.
- Made of borosilicate glass
With sealed-in fritted glass disc, they are available in coarse (C) and medium (M) porosities.
Recommended for use with LG-7820 heavy wall filter flasks.
Customers Who Bought This Also Bought