Soldering Gun
Catalog # 470228-924
Supplier: KIDDER MFG. CO. LTD
Specifications
- Description:Soldering gun
- Cat. no.:470228-924
Specifications
About this item
More comfortable style to hold than traditional design.
- Pistol grip
This soldering gun is 100 watts. 40 gm solder tube included.
Certifications: CSA approved.
Customers Who Bought This Also Bought