Smart® Assembled Respirator and Cartridge, Multi-Gas/Vapor, Moldex®
# de catalogue 56219-172
Fournisseur: Moldex/Metric
Small Business Enterprise
Spécifications
- Description:Smart® Cartridge
- Environmentally Preferable:Safer Human and Environmental Health
- Size:Medium
- Cat. no.:56219-172
- NIOSH Approval:TC-23C-1809
- Supplier No.:8602
Spécifications
A propos de cet article
Smart® Cartridge comes with a built-in inhalation valve, plus an easy snap-in mount
- 100% PVC-free
- Multi-gas/vapor Smart® cartridge simplifies inventory
- Approved for ten common gas/vapors
- Cartridge selection made simpler and misuse reduced
Use with N95 or P100 particulate filters against even more combinations of contaminants.
Customers Who Bought This Also Bought