Decapper for Vials, MicroSlov
# de catalogue 97054-996
Fournisseur: MicroSolv Technology Corp
Spécifications
- Description:Decapper Tool
- Cat. no.:97054-996
- Seal Size:20 mm
- Supplier No.:95025-DC
Spécifications
A propos de cet article
These decapper tools are used for detaching aluminum Headspace Caps.
Customers Who Bought This Also Bought