Tube Furnaces with Stand for Horizontal or Vertical Operation, Model RT, Nabertherm
Supplier: NABERTHERM INC.
Certificates
About this item
These compact tube furnaces are used when laboratory experiments must be performed horizontally, vertically, or at specific angles.
- Tmax 1100, 1300 or 1500 °C
- Compact design
- Vertical or horizontal operation freely adjustable
- Working height freely adjustable
- Working tube made of C 530 ceramic
- Type S thermocouple
- Operation also possible separate from stand if safety guidelines are observed
- Control system integrated in furnace base
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
The ability to configure the angle of tilt and the working height, and their compact design, also make these tube furnaces suitable for integration into existing process systems.
BodyTextAccessories: Optional accessories include the gas supply system 1, and charge control. Contact your Avantor representative for more information.
Ordering information: These tube furnaces can be ordered in a variety of heated lengths, maximum temperatures, and voltage options. Models rated up to 1100, 1300 or 1500 °C. Control options include the B510 controller (5 programs with 4 segments each) or C550 controller (10 programs with 20 segments each).
Customers Who Bought This Also Bought