VWR® 20 mm Crimp Seal Headspace Vials
Supplier: VWR International
Certificates
About this item
Clear 20 ml headspace vials designed to fit most headspace autosamplers.
- Round bottom vials fit PerkinElmer®, Tekmar®, and Varian® headspace autosamplers
- Flat bottom vials fit Agilent® and Carlo Erba autosamplers
Ordering information: 20 mm crimp seals have to be ordered separately.
Customers Who Bought This Also Bought