SP Wilmad-LabGlass Aluminum Bench Top Hemi Heating Mantles, SP Industries
Supplier: SP Industries
Certificates
About this item
These benchtop heating mantles are used for round bottom glass flasks.
- Hemispherical
With an outside aluminum housing and an internal glass fabric covering, these units can be used at temperatures up to 450°C and are available for flask sizes up to 22,000 mL. 115V.
Ordering information: Supplied with detachable 4 foot, 3-wire type SJT cord, and three wire terminal with one wire grounded to the housing.
Customers Who Bought This Also Bought