Neoprene Gloves, Mapa Professional
Fournisseur: MAPA
Certificats
A propos de cet article
Chemical resistant neoprene gloves with pinked cuff type and Z pattern finish.
- Embossed texture for better grip on wet objects
- Added comfort and thermal insulation, due to the cotton-knit lining
Spécifications
- Catalog No:
- 75796-100
- 75796-094
- 75796-092
- 75796-098
- 75796-096
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- Finish:
- Z-Grip (embossed)
- Z-Grip (embossed)
- Z-Grip (embossed)
- Z-Grip (embossed)
- Z-Grip (embossed)
- Cuff style:
- Pinked
- Pinked
- Pinked
- Pinked
- Pinked