Polyester/Cellulose Cleanroom Wipers, FG Cleanwipes
# de catalogue 89096-118
Fournisseur: Lymtech
Spécifications
- Dimensions:30.5 x 30.5 cm (12 x 12")
- Cat. no.:89096-118
- Packaging:150 Wipers/Bag, 10 Bags/Case
- Supplier No.:7-C30-BBL-00
Spécifications
A propos de cet article
These cleanroom wipers are made from a hydro-entangled blend of 55% cellulose and 45% polyester.
- Super absorbent
- Extremely low particle generation
- No chemical binders
Recommended for ISO 5-8 controlled environments.
Customers Who Bought This Also Bought