High-Temperature Furnaces with SiC Rod Heating, Up to 1600 °C, Nabertherm
Fournisseur: Avantor
Certificats
A propos de cet article
These powerful laboratory muffle furnaces are available for temperatures up to 1400, 1500, 1550 or 1600 °C.
- Tmax 1400, 1500, 1550 or 1600 °C
- Working temperature 1500 °C (for high-temperature furnaces LHTC ../16), increased wear and tear must be expected in case of working at higher temperatures
- Furnace chamber lined with first-class, long-life fiber material, unclassified
- Housing made of sheets of textured stainless steel
- Dual shell housing for low external temperatures and high stability
- Optional fold down door (LHTC) which can be used as work platform or lift up door (LHTCT) with hot surface facing away from the Operator. High-temperature furnace LHTCT 01/16 only with lift up door)
- Switching system with solid-state-relays, power tuned to the SiC rods
- Easy replacement of heating rods
- Adjustable air inlet opening, exhaust air opening in the roof
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
The durability of the SiC rods in periodic use, in combination with their high heating speed, make these high-temperature furnaces to all-rounders in the laboratory. Heating times of 40 minutes to 1400 °C can be achieved, depending on the furnace model and the conditions of use.
BodyTextAccessories: Optional accessories include over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; square saggar for charging of up to three layers; lid for top saggar; manual or automatic gas supply system; process control and documentation via VCD software package for monitoring, documentation and control. Contact your VWR representative for more information.
Ordering information: These high-temperature furnaces with SiC rod heating can be ordered with a variety of sizes, temperature, and voltage options. Models are rated up to 1400, 1500, or 1600 °C. Control the furnaces with the C550 controller (10 programs with 20 segments each).
Customers Who Bought This Also Bought