High-Temperature Furnaces with MoSi₂ Heating Elements, Up to 1800 °C, LHT Model, Nabertherm
Fournisseur: NABERTHERM INC.
Certificats
A propos de cet article
Designed as tabletop models, these compact high-temperature furnaces have a variety of advantages.
- Tmax 1600, 1750, or 1800 °C
- High-quality molybdenum disilicide heating elements
- Furnace chamber lined with first-class, long-life fiber material, unclassified
- Housing made of sheets of textured stainless steel
- Dual shell housing with additional fan cooling for low surface temperature
- Compact design with lift door, opening upwards
- Adjustable air inlet
- Exhaust air opening in the roof
- Type B thermocouple
- Switchgear with steady control of the heating elements
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
The first-class workmanship using high-quality materials, combined with ease of operation, make these furnaces all-rounders in research and the laboratory. These high-temperature furnaces are also perfectly suited for the sintering of technical ceramics, such as zirconium oxide dental bridges.
BodyTextAccessories: Optional accessories include over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; square saggar for charging of up to three layers; protective gas connection to purge with non-flammable protective or reaction gases; manual or automatic gas supply system; process control and documentation via VCD software package for monitoring, documentation and control. Contact your Avantor representative for more information.
Ordering information: These high-temperature furnaces with MoSi2 heating elements can be ordered with a variety of sizes, temperature, and voltage options. Models are rated up to 1600, 1650, 1750 or 1800 °C. Furnaces come with the P570 controller (50 programs with 40 segments each).
Customers Who Bought This Also Bought