ContactUsLinkComponent
8330 Calibration Mix #2 (7 components), Restek
8330 Calibration Mix #2 (7 components), Restek
# de catalogue RK31451
Fournisseur:  Restek
Numéro CAS:  
SV MIX 8330 CAL STD 2
8330 Calibration Mix #2 (7 components), Restek
# de catalogue RK31451
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Concentration:
    1000 µg/ml each in acetonitrile
  • Description:
    8330 Calibration Mix #2
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK31451
  • Certification:
    CRM
  • Supplier No.:
    31451
Spécifications
A propos de cet article
EPA method 8330 is used to measure explosives residues in water and soil samples, using HPLC with UV detection. Target analytes are nitroaromatic and nitramine explosives and their degradation products. Obtaining pure, neat compounds for Method 8330 reference standards can be very difficult. To ensure the highest quality standards, Restek's Quality Control (QC) lab confirms the identity and purity of mixture components and solvents using one or more of the following techniques: GC-FID, HPLC, GC-ECD, GC-MS, LC-MS, refractive index, and melting point.
8330 calibration mix #2 contains 2-amino-4,6-dinitrotoluene, 4-amino-2,6-dinitrotoluene, 2,6-dinitrotoluene, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, tetryl.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Customers Who Bought This Also Bought