Autoclave Bag Holders, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Epoxy-coated, wire-framed bag holders keep biohazard bags open and ready for use.
- Separate hooked ring rests over the top to keep the bag in place
- Circular shape keeps the bag wide open for easy access
Simply place a biohazard bag into the sturdy wire holder and stretch the bag over the top ring to secure it in place.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.