Bronze Phase Separator, Airgas®
# de catalogue 47752-624
Fournisseur: Airgas
Spécifications
- Length:2"
- Description:Phase separator, nitrogen, argon and helium usage
- Material:Bronze
- Cat. no.:47752-624
- Supplier No.:Y15-PSB38
- Inlet Connection:⅜" FNPT