SP Wilmad-LabGlass Mobile Phase Filtration/Degassing Cap, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Filtration/Degassing caps are constructed of PTFE with polypropylene vacuum adapter for 1/4" I.D
Lower screw cap has 45 mm thread and upper screw clamp holds mobile phase pickup adapter. Caps include a removable coarse fritted glass filter support (specially treated to resist flaking) and a Viton® O-ring for sealing to the reservoir.
tubing.
Customers Who Bought This Also Bought