ContactUsLinkComponent
Pierce ProFound™ PolyHis-Protein Interaction Pull Down Kit, Thermo Scientific
Pierce ProFound™ PolyHis-Protein Interaction Pull Down Kit, Thermo Scientific
# de catalogue CAPI21277
Fournisseur:  Thermo Scientific
Numéro CAS:  
Kit pierce His Protein Interaction Pull-Down 25rxn
Pierce ProFound™ PolyHis-Protein Interaction Pull Down Kit, Thermo Scientific
# de catalogue CAPI21277
Fournisseur:  Thermo Scientific
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    PolyHis-Protein:Protein Interaction Kit
  • No. of Reactions:
    25 Pull-down reactions, each using 25 µl of resin
  • Supplier No.:
    21277
Spécifications
A propos de cet article
The Pierce™ His Protein Interaction Pull-Down Kit contains the necessary components to capture and purify proteins that interact with His-tagged fusion proteins.
  • Complete kits provide all components and detailed protocols for purifying protein:protein interactions
  • No special equipment needed – use common laboratory equipment and reagents
  • Microcentrifuge spin columns facilitate simple and efficient manipulation of agarose beads, including simple processing of multiple samples
  • Instructions include protocols for use with bait and prey proteins expressed from a variety of sample types
  • Cobalt chelate resin is more specific for histidine-tagged fusion proteins than nickel resins, resulting in less non-specific binding
  • Binds 10 to 25 mg of histidine-tagged fusion protein per mL of resin
Applications include discover a new protein:protein interaction from a cell lysate; confirm a putative interaction from a cell lysate or with a previously purified protein; extract protein:protein interaction information from in vitro transcription/translation lysates.
Delivery: Complete kit includes Cobalt chelate Agarose resin, (750 µL); Pull-Down Lysis Buffer, (250 mL); Imidazole solution (4 M), (5 mL); TBS pack (makes 500 mL); Spin columns and accessories, 27 columns; and Microcentrifuge collection tubes, 100 tubes.
Customers Who Bought This Also Bought