WelchNet™ Modular Lab Vacuum Network Accessories, Welch Vacuum
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
A powerful alternative to house vacuum; minimizes cross contamination and is easily adapted as needs change.
- PTFE and PVDF connections wetted materials offer chemical resistance
Choose WelchNet fittings and tubing to ensure all modular vacuum network components interface properly.
Use fittings and tubing to customize your vacuum network to your particular vacuum needs.
Customers Who Bought This Also Bought