WelchNet™ Modular Lab Vacuum Network Pumps and Controllers, Welch Vacuum
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
WelchNet is a powerful alternative to house vacuum; minimizes cross contamination and is easily adapted as needs change
- Quick and simple to install a modular lab network
- Resistant to chemical attack
- Provides stable vacuum
- Expands easily as needs change
Quick and easy automated, digital vacuum setting and control. Controls vacuum level, cooling water, and venting. Choose LVS systems to combine vacuum controller and vacuum pump with catchpot. Choose standalone vacuum controllers for control of independent vacuum pump.
Controllers and LVS systems control vacuum levels.
Customers Who Bought This Also Bought