Premium Brilliant Series Heavy Duty Vest, ML Kishigo
Fournisseur: M.L. KISHIGO
Certificats
A propos de cet article
This vest will keep wearers more visible with the extra reflective trim.
- Ultra-cool 100% polyester mesh
- Brilliant Trim reflective edging improves low light visibility
- Zipper front closure reinforced with durable webbing
- 2" wide reflective material with 3" breathable contrasting color
- Left and right chest mic tabs
- 4 outside and 2 inside pockets
- Available in Lime and Orange
ML Kishigo's ANSI Class 2 Premium Brilliant Series Heavy Duty Vest features reflective trim to improve low light visibility. This vest has 6 pockets and zipper front closure.
Certifications: Meets ANSI 107 Class 2 requirements.
Spécifications
- Catalog No:
- 75998-128
- 75998-172
- 75998-170
- 75998-126
- 75998-168
- 75998-166
- 75998-180
- 75998-178
- 75998-176
- 75998-174
- 75998-276
- 75998-274
- 75998-280
- 75998-278
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester