ContactUsLinkComponent
SP Wilmad-LabGlass Polypropylene Reservoir Storage Cap, SP Industries
SP Wilmad-LabGlass Polypropylene Reservoir Storage Cap, SP Industries
Catalog # 14212-422
Supplier:  SP Industries
CAP RED GL-45 W/ PTFE LINER
SP Wilmad-LabGlass Polypropylene Reservoir Storage Cap, SP Industries
Catalog # 14212-422
Supplier:  SP Industries
Specifications
  • Description:
    Polypropylene Reservoir Storage Cap
  • Cat. no.:
    14212-422
  • Supplier No.:
    BP-1909-000
Specifications
About this item
This resevoir storage cap is red, made from polypropylene, and features a 45 mm thread and PTFE liner.
Fits all CHROMWARE® HPLC reservoirs.
Customers Who Bought This Also Bought