ContactUsLinkComponent
Elephant Toothpaste
Elephant Toothpaste
# de catalogue 470228-174
Fournisseur:  Ward's Science
Elephant Toothpaste Kit For grades 2-5
Elephant Toothpaste
# de catalogue 470228-174
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Elephant Toothpaste
  • No. of Lab Groups:
    1
  • Cat. no.:
    470228-174
Spécifications
A propos de cet article
Exciting SAFE Demo for Young Scientists.
  • Aligned to Next Generation science standards (NGSS)
  • Developed for grades 2-5
Check out this fun demonstration that is classroom safe and shows the effects of a catalyst on a chemical reaction. Discussion topics can include basic chemical reactions, phase changes and catalysts.
Ordering information: Includes 150 mL Hydrogen Peroxide 3%, 30 mL Dish Soap, 1 Clear Bottle (8oz), 5 g Yeast, 5 Plastic Cups (5oz), 5 Stir Sticks.
Caution: WARNING CHOKING HAZARD SMALL PART. Not for children under 3 yrs.
Customers Who Bought This Also Bought