Latent Fingerprint Powder
Fournisseur: FORENSICS SOURCE
Certificats
A propos de cet article
A variety of powders ensure that you are prepared for collecting evidence in any field condition.
- Black powder works for general applications
- Silver-Gray powder designed for glass, metallic, varnished or rubber surfaces
- Fluorescent powder combines with ultra violet or laser light sources
- Size: two ounces
The common black powder works for most general applications and the bi-chromatic powder highlights fingerprints on light or dark surfaces.
For glass, metallic, varnished, or hard rubber surfaces, the highly sensitive silver-gray powder reveals clear, accurate prints. When combined with ultraviolet or laser light sources, the fluorescent powder shows detailed fingerprints.
Customers Who Bought This Also Bought