ContactUsLinkComponent
Soap Power!
Soap Power!
# de catalogue 470228-172
Fournisseur:  Ward's Science
Soap Power Kit For Grades 2-5
Soap Power!
# de catalogue 470228-172
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Soap Powder!
  • No. of Lab Groups:
    6
  • Cat. no.:
    470228-172
  • Includes:
    Teacher Manual and Student Worksheets
Spécifications
A propos de cet article
Fun with Surface Tension
  • Aligned to Next Generation science standards (NGSS)
  • Developed for grades 2-5
Why are raindrops the shape that they are? How do detergents work? Simply put - surface tension. This kit contains materials to demonstrate the concept of surface tension to your classroom using soap as a power source! Kit contains enough materials for 6 groups of 4.
Ordering information: Includes 6 x 5 mL Dropper Bottle Liquid Dish Soap, 2 Foam Trays, 6 Plastic Trays, 6 pepper Packets.
Caution: WARNING CHOKING HAZARD SMALL PART. Not for children under 3 yrs.
Customers Who Bought This Also Bought