Wards® Nutrient Agar Powdered Media
Fournisseur: Ward's Science
Certificats
A propos de cet article
High quality microbiological media at an economical price.
- SDS forms available for each media type
- Carefully blended for consistency and reliability
- Longer shelf life than prepared media
- Economical alternative
Use's Ward's® own dehydrated media for many of your routine microbiology applications.
Combine powder with water according to the mixture instructions attached on the packaging label and sterilize for ready-to-use bacterial media.
Customers Who Bought This Also Bought