V-300 Vacuum Pump with I-300 Controller, BUCHI
Supplier: Avantor
Certificates
About this item
The chemically resistant PTFE diaphragm vacuum pump impresses with its quiet and economical operation
- Quiet operation
- Oil-free PTFE diaphragm pump
- Reliable
- Visible operating membranes
- Speed controlled
The V-300 vacuum pump works quietly at 32 dB, with PTFE membranes visible to monitor presence of any condensed solvent inside the pump. The V-300 pump is controlled by the I-300 interface and is optimally suited for use with the R-300 Rotavapor®.
It is optimally suited to be combined with the Rotavapor® R-300 and I-300 Controller.
Certifications: CE, CSA and CUS compliant.
Ordering information: All accessories and connections are included for out of the box usage. Waste level sensors are sold separately. Please contact your Avantor representative for more information. Additional accessories are required for adaptation with older BUCHI systems. Please contact your Avantor representative for further assistance.
Specifications
- Catalog No:
- 10023-982
- 10023-984
- CA10023-984
- CA10023-982
- CA10023-978
- Final vacuum:
- 5 mbar ±2 mbar
- 5 mbar ±2 mbar
- 5 mbar ±2 mbar
- 5 mbar ±2 mbar
- 5 mbar ±2 mbar
- Height:
- 321 mm
- 321 mm
- 321 mm
- 321 mm
- 321 mm
- Length:
- 200 mm
- 200 mm
- 200 mm
- 200 mm
- 200 mm
- Suction capacity:
- 1.8 m³/h (vacuum)
- 1.8 m³/h (vacuum)
- 1.8 m³/h (vacuum)
- 1.8 m³/h (vacuum)
- 1.8 m³/h (vacuum)
- Weight:
- 7.6 kg
- 7.6 kg
- 7.6 kg
- 7.6 kg
- 7.6 kg
- Width:
- 291 mm
- 291 mm
- 291 mm
- 291 mm
- 291 mm