Total Sugar Test Strips, EMD Millipore
Fournisseur: MilliporeSigma
Certificats
A propos de cet article
Total Sugar Test can be used to monitor glucose and fructose in food, such as potatoes, and in fruit juices, and also optimize your wine making process.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.