Specific Gravity/Heat Set
# de catalogue 470227-640
Fournisseur: Wards
Spécifications
- Description:Specific Gravity/Heat Set
- Cat. no.:470227-640
Spécifications
A propos de cet article
A useful set for density, specific heat and gravity.
- Useful in thermodynamics and density studies
- Includes instruction manuals
- Hooks allow easy measurements from a spring scale
Help beginning students conceptually understand density. A cylinder of zinc, copper, aluminum, tin, and steel, each machine stamped with identification, are included in the kit along with a complete instruction manual that gives background on density, specific gravity, and specific heat and contains three inquiry based student activities with capture sheets. Each cylinder has the same 1/2” diameter and mass but a different length.
Customers Who Bought This Also Bought