Eye and Face Shower, Galvanized Pipe, Encon® Safety Products
Fournisseur: Encon Safety Products
Certificats
A propos de cet article
This combination shower/face and eye wash with bowl is made with schedule 80 hot dip galvanized steel piping.
- ¹/₂"CP bronze EFW valve
- 1" CP bronze shower stay open valve with CP ball
- Stainless steel valve stem
- Stainless steel pull road assembly with actuation graphic
Eyewash fountain have two choices that is ABS plastic heads with float off covers and acetal heads with hinged covers.
Designed with varying hydraulic conditions from 30 to 70 psi (206.8 to 482.6 kPa).
Facewash fountain is offers with ABS plastic heads with wye and float off covers.
Customers Who Bought This Also Bought